Learn More
Electron Microscopy Sciences Microtube 80-Well Microtube Rack, Yellow
SDP
Catalog No. 501899023
Encompass_Preferred
Popular fraction-collector-plate style rack. Rack holds 1.5 to 2.0 ml microtubes in a 5 x 16 array. Tubes sit in large wells with conical bottoms. Polypropylene rack is ideal for freezer storage. Autoclavable. Rack measures 8.875 x 2.625 x 1.125".
Color: Yellow
Catalog No. 50-189-9023 Supplier Electron Microscopy Sciences Supplier No. 61603Y
Popular fraction-collector-plate style rack. Rack holds 1.5 to 2.0 ml microtubes in a 5 x 16 array. Tubes sit in large wells with conical bottoms. Polypropylene rack is ideal for freezer storage. Autoclavable. Rack measures 8.875 x 2.625 x 1.125".
Color: Yellow