Learn More
Analytical Sales & Services MicroTee for .025" Tubing Sleeves
Catalog No. 5000804562
Encompass
Micro Tee Assembly for .025" Microtight Sleeves .006" Thru-Hole Swept Volume 29nL Includes (3) 55172 and (3) 59416 Fittings
Qty/Pkg: EACH
Catalog No. 50-008-04562 Supplier Analytical Sales & Services Supplier No. 59775
Micro Tee Assembly for .025" Microtight Sleeves .006" Thru-Hole Swept Volume 29nL Includes (3) 55172 and (3) 59416 Fittings
Qty/Pkg: EACH