Learn More
Analytical Sales & Services MicroTee for 360µm OD Tubing
Catalog No. 5000804508
Encompass
MicroTee for 360µm OD Tubing .006" Thru Hole Swept Volume 29nL Includes (3) 55152 and (3) 59416BLK Fittings
Qty/Pkg: EACH
Catalog No. 50-008-04508 Supplier Analytical Sales & Services Supplier No. 59888
MicroTee for 360µm OD Tubing .006" Thru Hole Swept Volume 29nL Includes (3) 55152 and (3) 59416BLK Fittings
Qty/Pkg: EACH