Learn More
IDEX Health & Science Upchurch MicroTee PEEK - 1/16" w/Fittings
Catalog No. NC1735973
Encompass_Preferred
Biocompatible, MicroTee is used to effectively join 1/16" capillary tubing together. It is specifically designed with a 0.006" thruhole that delivers a low swept volume. It is made from inert PEEK, which is ideal for chemical resistance. It includes (3) F-132 and (3) P-416 Fittings.
Catalog No. NC1735973 Supplier IDEX Health & Science Upchurch Supplier No. P890
Biocompatible, MicroTee is used to effectively join 1/16" capillary tubing together. It is specifically designed with a 0.006" thruhole that delivers a low swept volume. It is made from inert PEEK, which is ideal for chemical resistance. It includes (3) F-132 and (3) P-416 Fittings.