Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ MicroTest™ Tubes

Catalog No. r12533
Change view
Click to view available options
Closure Color:
Green
Red
Media Type:
M5
M6, 1.5 mL
M6, 3 mL
Includes:
Glass Beads
Quantity:
72 Tubes/Pk.
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Closure Color Media Type Includes Quantity
R12533 Red M6, 3 mL Glass Beads 72 Tubes/Pk.
17-077-913 Green M5 Glass Beads 72 Tubes/Pk.
R12530 Red M6, 1.5 mL Glass Beads 72 Tubes/Pk.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. R12533 Supplier Thermo Scientific™ Supplier No. R12533
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Maintain viability of viruses chlamydiae, mycoplasma and urealasma with top quality transport and storage media that comes in four versatile formulations.

Maintain organism viability and counts for verification studies, quality control procedures, and repeat testing of viruses, chlamydiae, mycoplasma and ureaplasma with top quality media from the experts in media. Thermo Scientific™ MicroTest™ Tubes come in four media formulations to offer flexibility in both transport and storage.

Designed for the collection, transportation, maintenance and long-term storage of viral specimens, chlamydiae, mycoplasma and ureaplasma, Thermo Scientific MicroTest products are top quality. From formulation to packaging, MicroTest products maintain the viability of organisms through freeze-thaw cycles while inhibiting antimicrobial contaminants and promoting safety.

MicroTest™ M4™ contains gelatin, vancomycin, amphotericin B, and colistin for the transport of viruses, Chlamydiae, Ureaplasmas, and Mycoplasmas.

MicroTest™ M4RT™ contains gelatin, gentamicin, and amphotericin B for the transport of viruses and Chlamydiae. MicroTest™ M5™ contains vancomycin, amphotericin B, and colistin and protein stabilizers for the transport of viruses, Chlamydiae, Ureaplasmas, and Mycoplasmas.

MicroTest™ M6™ contains gelatin, vancomycin, amphotericin B, and colistin for the transport of viruses, Chlamydiae, Ureaplasmas, and Mycoplasmas.

MicroTest Tubes come with 1.5mL or 3mL of liquid medium in a 15mL conical tube with 3 glass beads.

Keep your lab moving in the right direction with a dependable set of solutions for transport, holding, maintenance and long-term culture storage with the MicroTest family of collection and transport media.

Trusted reliable results
  • Stringent quality control, including cellular toxicity on each lot number
  • Patented formulations with stabilizing proteins, buffers, and specific antibiotics
Compatibility and viability
  • Provides compatible transport for cell culture, enzyme immunoassays, and molecular diagnostic tests
  • Maintains organism viability and counts for better, more reliable results during validation/verification studies, quality control procedures, and repeat testing
Convenient, accommodating format
  • 1.5mL and 3.0mL fill volumes allow multiple tests from one specimen
  • Individual transport media tubes or complete kits offer flexibility and versatility
Safe, sturdy packaging
  • Tough, break-resistant tubes with secure, color-coded screw caps prevent leaking and exposure to infectious agents
  • Kits include leak-resistant polypropylene bag for safe sample transport

Not all products are available for sale in all territories.

Please inquire. Remel™ and Oxoid™ products are now part of the Thermo Scientific brand, combining powerful manual, semi-automated and fully automated test products and a comprehensive line of media and diagnostic products to offer a complete, end-to-end solution to quickly deliver the products you need and the quality results your laboratory depends on.
TRUSTED_SUSTAINABILITY

Specifications

Product Type Transport Tube
Format Tube
Media Type M6, 3 mL
Closure Color Red
Compatibility Culture, Shell vial, Freeze-thaw cycles, Culture maintenance, ELISA, PCR, DNA probe technology
Description M6 Tube
For Use With (Application) Viruses: Chlamydiae, Ureaplasmas, and Mycoplasmas
Includes Glass Beads
Packaging Packaged in partitioned boxes.
Quantity 72 Tubes/Pk.
Storage Requirements 2°C to 30°C
Transport Temperature 2° to 30°C
Volume (Metric) 3 mL
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.