Learn More
IDEX Health & Science Upchurch MICROTIGHT ADAPTER FOR TUBING
Catalog No. NC0789877
Encompass_Preferred
These special MicroTight Adapters were created to facilitate capillary tubing connections to standard 1/16 in. or 1/32 in.OD tubing. Our Adapters create true zero dead volume (ZDV) which allows tubing to butt against each other for an uninterrupted fluidic path. Includes (1)1 F-120R, (1) F-126S, (1) P-554.
Catalog No. NC0789877 Supplier IDEX Health & Science Upchurch Supplier No. P881
These special MicroTight Adapters were created to facilitate capillary tubing connections to standard 1/16 in. or 1/32 in.OD tubing. Our Adapters create true zero dead volume (ZDV) which allows tubing to butt against each other for an uninterrupted fluidic path. Includes (1)1 F-120R, (1) F-126S, (1) P-554.