Learn More
Analytical Sales & Services MicroTight Union for 360µm OD Tubing
Catalog No. 5000804561
Encompass
PEEK Microtight Union 0.015" 360µm OD to 360µm OD Includes; (2) 55152 Micro Ferrules and (2)59416 Female Nuts No Sleeves Required for the Connections
Qty/Pkg: EACH
Catalog No. 50-008-04561 Supplier Analytical Sales & Services Supplier No. 59772
PEEK Microtight Union 0.015" 360µm OD to 360µm OD Includes; (2) 55152 Micro Ferrules and (2)59416 Female Nuts No Sleeves Required for the Connections
Qty/Pkg: EACH