Learn More
Sigma Aldrich Fine Chemicals Biosciences MicroTissuesR 3D Petri Dis
Supplier:  Sigma Aldrich Fine Chemicals Biosciences Z7640196EA
Encompass
When the gelled agarose is removed from the micro-mold it is transferred to a standard 12 well or 24 well tissue culture dish and equilibrated with cell culture medium.Since the agarose is transparent the spheroids or microtissues that form at the bottom of each agarose micro-well can be easily viewed using a standard inverted microscope using phase contrast bright field or fluorescence microscopy.The micro-molds are reusable up to 12 times and can be sterilized via a standard steam autoclave (30 min dry cycle)The micro-molds should be stored in a covered container to maintain sterility and to avoid collecting dust or fibres on their small featuresAny microscope with sufficient magnification and reSOLUTIONon functions for cell based views or images will be suitable for use with the 3D Petri Dish products
Catalog No. 50-290-5676