Learn More
Andwin Scientific MICROTITER FORMAT COMB FOR DNA
SDP
Catalog No. 50820196
Encompass_Preferred
Microtiter format Comb for DNA Plus;High Capacity Horizontal Gel System. Teeth/Comb: 25. Thickness: 1.0 mm. Tooth Width: 7.5 mm. Volume/Well: 22 ul.
Catalog No. 50-820-196 Supplier Andwin Scientific Supplier No. 34314496
Microtiter format Comb for DNA Plus;High Capacity Horizontal Gel System. Teeth/Comb: 25. Thickness: 1.0 mm. Tooth Width: 7.5 mm. Volume/Well: 22 ul.