Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Poltex Microtiter Plate Carrying Caddy - Five Slot
SDP

Catalog No. 22022547
Change view
Click to view available options
Holds:
Microplates
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Holds
22-022-547 Microplates
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 22-022-547 Supplier Poltex Supplier No. PLATRCK5
Get a Poltex Pipette Holder with $3,000 Purchase
View all promotions
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Convenient carrier for microtiter plates. Front door lifts off for loading, but remains securely in place for transport.

  • Front door lifts off for loading, but remains securely in place for transport
  • Durable PETG construction

Specifications

Autoclavable Not Autoclavable
Color Clear
Holds Microplates
Material Polycarbonate
Positions 1
Shape Vertical
Stackable No
For Use With (Equipment) Microtiter plate
Dimensions (L x W x H) 5.6 x 4.2 x 8 in.
Length (Metric) 142.24 mm
Product Type Microplate Rack
Height (Metric) 203.20 mm
Width (Metric) 106.68 mm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.