Learn More
Andwin Scientific MICROTITER SHARKTOOTH COMB FOR
SDP
Catalog No. 50820221
Encompass_Preferred
Microtiter Sharktooth comb for Sequencing Gel System, 35 x 45 cm. Teeth/Comb: 72. Thickness: 0.4 mm. Volume/Well: 10 ul.
Catalog No. 50-820-221 Supplier Andwin Scientific Supplier No. 34855472
Microtiter Sharktooth comb for Sequencing Gel System, 35 x 45 cm. Teeth/Comb: 72. Thickness: 0.4 mm. Volume/Well: 10 ul.