Learn More
Biolog Inc MicroPlate™ Trays w/ Lids | 25/pack
SDP
Catalog No. NC1382824
Encompass
Virgin polystyrene, 96 flat-bottom wells, half the volume of standard wells, low evaporation lids, gamma irradiated, trays & lids packaged separately
Catalog No. NC1382824 Supplier Biolog Inc Supplier No. 30311
Virgin polystyrene, 96 flat-bottom wells, half the volume of standard wells, low evaporation lids, gamma irradiated, trays & lids packaged separately