Learn More
Electron Microscopy Sciences Sturkey Infinity Low Profile Disposible Microtome Blades, Standard Dispenser
SDP
Catalog No. 501929181
Encompass_Preferred
These unique blades feature innovative edge coatings that improves durability, lubricity and longevity. Made in the US. The INFINITY features a triple faceted, 35 degree cutting edge on a 80mm length blade. It is designed to provide incredible longevity and performance for general paraffin and cryostat use. Coated with PTFE-like substance to allow a smooth cut.
Package: Standard Dispenser
Length: 80 mm
Angle: 35°
This item is not returnable. View return policy
Catalog No. 50-192-9181 Supplier Electron Microscopy Sciences Supplier No. 6305901
These unique blades feature innovative edge coatings that improves durability, lubricity and longevity. Made in the US. The INFINITY features a triple faceted, 35 degree cutting edge on a 80mm length blade. It is designed to provide incredible longevity and performance for general paraffin and cryostat use. Coated with PTFE-like substance to allow a smooth cut.
Package: Standard Dispenser
Length: 80 mm
Angle: 35°