Learn More
Electron Microscopy Sciences Sturkey Monarch Low Profile Disposible Microtome Blades, Standard Dispenser
SDP
Catalog No. 501929183
Encompass_Preferred
These unique blades feature innovative edge coatings that improves durability, lubricity and longevity. Made in the US. MONARCH blades are fully coated with a 35 degree edge. It is designed for general, all-purpose cutting applications (including cryostats.) Low profile only.
Package: Standard Dispenser
Length: 80 mm
Angle: 35°
This item is not returnable. View return policy
Catalog No. 50-192-9183 Supplier Electron Microscopy Sciences Supplier No. 6305903
These unique blades feature innovative edge coatings that improves durability, lubricity and longevity. Made in the US. MONARCH blades are fully coated with a 35 degree edge. It is designed for general, all-purpose cutting applications (including cryostats.) Low profile only.
Package: Standard Dispenser
Length: 80 mm
Angle: 35°