Learn More
Electron Microscopy Sciences Sturkey Extremus High Profile Disposible Microtome Blades, Standard Dispenser
SDP
Catalog No. 501929194
Encompass_Preferred
Extremus microtome blades are uniquely coated to suit the widest variety of cutting condition and feature a unique combination of durability and precision to save time and money.
Package: Standard Dispenser
Length: 80 mm
Angle: 35°
This item is not returnable. View return policy
Catalog No. 50-192-9194 Supplier Electron Microscopy Sciences Supplier No. 6305915
Extremus microtome blades are uniquely coated to suit the widest variety of cutting condition and feature a unique combination of durability and precision to save time and money.
Package: Standard Dispenser
Length: 80 mm
Angle: 35°