Learn More
Electron Microscopy Sciences Sturkey Premium High Profile Disposible Microtome Blades, Bulk Dispenser
SDP
Catalog No. 501929206
Encompass_Preferred
PREMIUM disposable microtome blade edges are uncoated. They are very durable and economical. They are offered in heavy duty high profile only.
Package: Bulk Dispenser
Length: 80 mm
Angle: 35°
This item is not returnable. View return policy
Catalog No. 50-192-9206 Supplier Electron Microscopy Sciences Supplier No. 6305927
PREMIUM disposable microtome blade edges are uncoated. They are very durable and economical. They are offered in heavy duty high profile only.
Package: Bulk Dispenser
Length: 80 mm
Angle: 35°