Learn More
Electron Microscopy Sciences Sturkey Premium High Profile Disposible Microtome Blades, Box Packaging
SDP
Catalog No. 501929207
Encompass_Preferred
PREMIUM disposable microtome blade edges are uncoated. They are very durable and economical. They are offered in heavy duty high profile only.
Package: Box Packaging
Length: 80 mm
Angle: 35°
This item is not returnable. View return policy
Catalog No. 50-192-9207 Supplier Electron Microscopy Sciences Supplier No. 6305928
PREMIUM disposable microtome blade edges are uncoated. They are very durable and economical. They are offered in heavy duty high profile only.
Package: Box Packaging
Length: 80 mm
Angle: 35°