Learn More
Electron Microscopy Sciences Sturkey Premium High Profile Disposible Microtome Blades, Standard Dispenser
SDP
Catalog No. 501929205
Encompass_Preferred
PREMIUM disposable microtome blade edges are uncoated. They are very durable and economical. They are offered in heavy duty high profile only.
Package: Standard Dispenser
Length: 80 mm
Angle: 35°
This item is not returnable. View return policy
Catalog No. 50-192-9205 Supplier Electron Microscopy Sciences Supplier No. 6305926
PREMIUM disposable microtome blade edges are uncoated. They are very durable and economical. They are offered in heavy duty high profile only.
Package: Standard Dispenser
Length: 80 mm
Angle: 35°