Learn More
Electron Microscopy Sciences Sturkey Select High Profile Disposible Microtome Blades, Standard Dispenser
SDP
Catalog No. 501929185
Encompass_Preferred
These unique blades feature innovative edge coatings that improves durability, lubricity and longevity. Made in the US. SELECT disposable microtome blades features state of the art grinding and coating techniques designed to create an edge that can produce superior sections with every cut. It is ideal for paraffin applications and cryostat use.
Package: Standard Dispenser
Length: 80 mm
Angle: 35°
This item is not returnable. View return policy
Catalog No. 50-192-9185 Supplier Electron Microscopy Sciences Supplier No. 6305905
These unique blades feature innovative edge coatings that improves durability, lubricity and longevity. Made in the US. SELECT disposable microtome blades features state of the art grinding and coating techniques designed to create an edge that can produce superior sections with every cut. It is ideal for paraffin applications and cryostat use.
Package: Standard Dispenser
Length: 80 mm
Angle: 35°