Learn More
Electron Microscopy Sciences Disposable Microtome Blades High Profile, 3"L x 0.562"W, 0.012" thick
SDP
Catalog No. 5025536
Encompass_Preferred
These stainless steel disposable blades feature an edge that is the product of years of research. Repeated testing was performed by histologists to assure that SHUR/SHARP blades represent the ultimate in consistency for sectioning technology. Chatterless and compression free ribbons will become routine no matter what the tissue is. For use with holder #63050-HP; #63050-HPE, and all high profile blade holders, Uncoated.
Size: 3"L x 0.562"W
Thickness: 0.012"
Catalog No. 50-255-36 Supplier Electron Microscopy Sciences Supplier No. 6306201
These stainless steel disposable blades feature an edge that is the product of years of research. Repeated testing was performed by histologists to assure that SHUR/SHARP blades represent the ultimate in consistency for sectioning technology. Chatterless and compression free ribbons will become routine no matter what the tissue is. For use with holder #63050-HP; #63050-HPE, and all high profile blade holders, Uncoated.
Size: 3"L x 0.562"W
Thickness: 0.012"