Learn More
Electron Microscopy Sciences Disposable Microtome Blades High Profile PTFE, 3"L x 0.553"W, 0.012" thick
SDP
Catalog No. 5025533
Encompass_Preferred
These stainless steel disposable blades feature an edge that is the product of years of research. Repeated testing was performed by histologists to assure that SHUR/SHARP blades represent the ultimate in consistency for sectioning technology. Chatterless and compression free ribbons will become routine no matter what the tissue is. Disposable high profile microtome blades, coated with PTFE.
Size: 3"L x 0.553"W
Thickness: 0.012"
Catalog No. 50-255-33 Supplier Electron Microscopy Sciences Supplier No. 6306410
These stainless steel disposable blades feature an edge that is the product of years of research. Repeated testing was performed by histologists to assure that SHUR/SHARP blades represent the ultimate in consistency for sectioning technology. Chatterless and compression free ribbons will become routine no matter what the tissue is. Disposable high profile microtome blades, coated with PTFE.
Size: 3"L x 0.553"W
Thickness: 0.012"