Learn More
Electron Microscopy Sciences Sturkey Silver Low Profile Disposible Microtome Blades, Standard Dispenser
SDP
Catalog No. 501929186
Encompass_Preferred
These unique blades feature innovative edge coatings that improves durability, lubricity and longevity. Made in the US. Silver Microtome Blades are coated with a nitride and copolymer finish to provide smooth, wrinkle-free sections. They are durable, long-lasting and ideal for general paraffin and cryostat use.
Package: Standard Dispenser
Length: 80 mm
Angle: 35°
This item is not returnable. View return policy
Catalog No. 50-192-9186 Supplier Electron Microscopy Sciences Supplier No. 6305906
These unique blades feature innovative edge coatings that improves durability, lubricity and longevity. Made in the US. Silver Microtome Blades are coated with a nitride and copolymer finish to provide smooth, wrinkle-free sections. They are durable, long-lasting and ideal for general paraffin and cryostat use.
Package: Standard Dispenser
Length: 80 mm
Angle: 35°