Learn More
Electron Microscopy Sciences Sturkey Diamond Low Profile Disposible Microtome Blades, Standard Dispenser
SDP
Catalog No. 501929203
Encompass_Preferred
DIAMOND disposable microtome blades have an amorphous diamond-coated cutting edge. The toughest, most durable blade in this line of products. Especially effective on hard tissue and decalcified bone.
Package: Standard Dispenser
Length: 80 mm
Angle: 35°
This item is not returnable. View return policy
Catalog No. 50-192-9203 Supplier Electron Microscopy Sciences Supplier No. 6305924
DIAMOND disposable microtome blades have an amorphous diamond-coated cutting edge. The toughest, most durable blade in this line of products. Especially effective on hard tissue and decalcified bone.
Package: Standard Dispenser
Length: 80 mm
Angle: 35°