Learn More
Electron Microscopy Sciences Sturkey Extremus Low Profile Disposible Microtome Blades, Box Packaging
SDP
Catalog No. 501929192
Encompass_Preferred
These unique blades feature innovative edge coatings that improves durability, lubricity and longevity. Made in the US. Extremus microtome blades are uniquely coated to suit the widest variety of cutting condition and feature a unique combination of durability and precision to save time and money.
Package: Box Packaging
Length: 80 mm
Angle: 35°
This item is not returnable. View return policy
Catalog No. 50-192-9192 Supplier Electron Microscopy Sciences Supplier No. 6305913
These unique blades feature innovative edge coatings that improves durability, lubricity and longevity. Made in the US. Extremus microtome blades are uniquely coated to suit the widest variety of cutting condition and feature a unique combination of durability and precision to save time and money.
Package: Box Packaging
Length: 80 mm
Angle: 35°