Learn More
Electron Microscopy Sciences Sturkey Extremus Low Profile Disposible Microtome Blades, Bulk Dispenser
SDP
Catalog No. 501929193
Encompass_Preferred
These unique blades feature innovative edge coatings that improves durability, lubricity and longevity. Made in the US. Extremus microtome blades are uniquely coated to suit the widest variety of cutting condition and feature a unique combination of durability and precision to save time and money.
Package: Bulk Dispenser
Length: 80 mm
Angle: 35°
This item is not returnable. View return policy
Catalog No. 50-192-9193 Supplier Electron Microscopy Sciences Supplier No. 6305914
These unique blades feature innovative edge coatings that improves durability, lubricity and longevity. Made in the US. Extremus microtome blades are uniquely coated to suit the widest variety of cutting condition and feature a unique combination of durability and precision to save time and money.
Package: Bulk Dispenser
Length: 80 mm
Angle: 35°