Learn More
Electron Microscopy Sciences Sturkey Gold Low Profile Disposible Microtome Blades, Standard Dispenser
SDP
Catalog No. 501929197
Encompass_Preferred
GOLD disposable microtome blades are the ultimate blade for cutting all types of skin and hard tissue. It is a high-performance specialty blade and its durability and toughness make it the best solution for hard tissue.
Package: Standard Dispenser
Length: 80 mm
Angle: 35°
This item is not returnable. View return policy
Catalog No. 50-192-9197 Supplier Electron Microscopy Sciences Supplier No. 6305918
GOLD disposable microtome blades are the ultimate blade for cutting all types of skin and hard tissue. It is a high-performance specialty blade and its durability and toughness make it the best solution for hard tissue.
Package: Standard Dispenser
Length: 80 mm
Angle: 35°