Learn More
Electron Microscopy Sciences Aluminium Microtome Chuck with Spigot, 26 mm diameter
SDP
Catalog No. 501930527
Encompass_Preferred
Aluminum Microtome Chucks with grooved surfaces for good specimen adhesion. Chucks have a 9.4 mm spigot.
Diameter: 26 mm
Catalog No. 50-193-0527 Supplier Electron Microscopy Sciences Supplier No. EMS066
Aluminum Microtome Chucks with grooved surfaces for good specimen adhesion. Chucks have a 9.4 mm spigot.
Diameter: 26 mm