Learn More
Electron Microscopy Sciences Aluminium Microtome Chuck with Spigot, 45 mm diameter
SDP
Catalog No. 501930526
Encompass_Preferred
Aluminum Microtome Chucks with grooved surfaces for good specimen adhesion. Chucks have a 9.4 mm spigot.
Diameter: 45 mm
Catalog No. 50-193-0526 Supplier Electron Microscopy Sciences Supplier No. EMS065
Aluminum Microtome Chucks with grooved surfaces for good specimen adhesion. Chucks have a 9.4 mm spigot.
Diameter: 45 mm