Learn More
Ted Pella Inc MICROTOME LUBRICATIONS KIT
SDP
Catalog No. NC9292163
Encompass
Tissue-Tek Microtome Lubrication Kit, Kit contains special low-Temperature lubricants necessary for routine maintenance of the Tissue-Tek Cryostat Microtomes, Includes Oil A, Oil B and Grease G, 1/EA
Catalog No. NC9292163 Supplier Ted Pella Inc Supplier No. 27244
Tissue-Tek Microtome Lubrication Kit, Kit contains special low-Temperature lubricants necessary for routine maintenance of the Tissue-Tek Cryostat Microtomes, Includes Oil A, Oil B and Grease G, 1/EA