Learn More
Electron Microscopy Sciences Microtome Oil 50 ML
SDP
Catalog No. 50996278
Encompass_Preferred
A chemically pure, high-grade lubricant used in precision instruments. Should be used periodically to lubricate cross-rollerbearings for microtomes.
Catalog No. 50-996-278 Supplier Electron Microscopy Sciences Supplier No. 60744
A chemically pure, high-grade lubricant used in precision instruments. Should be used periodically to lubricate cross-rollerbearings for microtomes.