Learn More
Electron Microscopy Sciences Crystallography MicroTool Kit 1, 20 Tools
SDP
Catalog No. 509053314
Encompass
These patent-pending MicroTools have tips used for protein crystals, single cells and manipulating small samples are made from soft, flexible microfabricated polymer films. The curvature of the tips gives them rigidity, but they can still easily be flexed. Tools designed for use with samples from a few micrometers to millimeter in size. These tools will cause far less  damage to fragile samples than metal tools, and they are optically and x-ray transparent. Each tool is mounted on a 0.025"/0.64 mm diameter non-magnetic stainless steel base.  The base is fitted in a handle 0.7 mm diameter, included in each kit. 20 micro tools for sample manipulating and measurements. Small, medium and large MicroSpoon, microchisel, MicroLasso, and MicroSieve, and MicroGripper, MicroRuler and two MicroSaws.
Catalog No. 50-905-3314 Supplier Electron Microscopy Sciences Supplier No. 6010001
These patent-pending MicroTools have tips used for protein crystals, single cells and manipulating small samples are made from soft, flexible microfabricated polymer films. The curvature of the tips gives them rigidity, but they can still easily be flexed. Tools designed for use with samples from a few micrometers to millimeter in size. These tools will cause far less  damage to fragile samples than metal tools, and they are optically and x-ray transparent. Each tool is mounted on a 0.025"/0.64 mm diameter non-magnetic stainless steel base.  The base is fitted in a handle 0.7 mm diameter, included in each kit. 20 micro tools for sample manipulating and measurements. Small, medium and large MicroSpoon, microchisel, MicroLasso, and MicroSieve, and MicroGripper, MicroRuler and two MicroSaws.