Learn More
Electron Microscopy Sciences Crystallography MicroTool Kit 3, 20 Tools
SDP
Supplier:  Electron Microscopy Sciences 6010003
Encompass
These patent-pending MicroTools have tips used for protein crystals, single cells and manipulating small samples are made from soft, flexible microfabricated polymer films. The curvature of the tips gives them rigidity, but they can still easily be flexed. Tools designed for use with samples from a few micrometers to millimeter in size. These tools will cause far less  damage to fragile samples than metal tools, and they are optically and x-ray transparent. Each tool is mounted on a 0.025"/0.64 mm diameter non-magnetic stainless steel base.  The base is fitted in a handle 0.7 mm diameter, included in each kit. 20 micro tools for sample measurement to accurately measure small and fragile samples with minimal change of sample damage. Tool tips are 10 um thick and are both optically and x-ray transparent.
Catalog No. 50-905-3316