Learn More
Electron Microscopy Sciences SureSeal S™ Microtubes, Cap and Stop-Pop Clips 1.5ml, Red
SDP
Catalog No. 501929288
Encompass_Preferred
Our SureSeal S microcentrifuge tubes are designed for a broad range of research applications, including enzyme digests, DNA isolation/purification, centrifugation, incubation and general sample/reagent storage. With a variety of design advances, SureSeal S tubes improve sample visualization, safety, stability and overall ease of use.
They are manufactured under clean room conditions, using only BioClear co-polymer, an advanced polypropylene that maximizes clarity. RNase, DNase and pyrogen free.
Color: Red
This item is not returnable. View return policy
Catalog No. 50-192-9288 Supplier Electron Microscopy Sciences Supplier No. 6302704
Our SureSeal S microcentrifuge tubes are designed for a broad range of research applications, including enzyme digests, DNA isolation/purification, centrifugation, incubation and general sample/reagent storage. With a variety of design advances, SureSeal S tubes improve sample visualization, safety, stability and overall ease of use.
They are manufactured under clean room conditions, using only BioClear co-polymer, an advanced polypropylene that maximizes clarity. RNase, DNase and pyrogen free.
Color: Red