Learn More
Electron Microscopy Sciences SureSeal S™ Microtubes with Cap and Stop-Pop Clips 2.0ml, Clear
SDP
Catalog No. 501929294
Encompass_Preferred
Our SureSeal S microcentrifuge tubes are designed for a broad range of research applications, including enzyme digests, DNA isolation/purification, centrifugation, incubation and general sample/reagent storage. With a variety of design advances, SureSeal S tubes improve sample visualization, safety, stability and overall ease of use.
They are manufactured under clean room conditions, using only BioClear co-polymer, an advanced polypropylene that maximizes clarity. RNase, DNase and pyrogen free.
Color: Clear
This item is not returnable. View return policy
Catalog No. 50-192-9294 Supplier Electron Microscopy Sciences Supplier No. 6302710
Our SureSeal S microcentrifuge tubes are designed for a broad range of research applications, including enzyme digests, DNA isolation/purification, centrifugation, incubation and general sample/reagent storage. With a variety of design advances, SureSeal S tubes improve sample visualization, safety, stability and overall ease of use.
They are manufactured under clean room conditions, using only BioClear co-polymer, an advanced polypropylene that maximizes clarity. RNase, DNase and pyrogen free.
Color: Clear