Learn More
Andwin Scientific MICROTUBE .5ML PP 1000C
SDP
Catalog No. NC0856993
Encompass_Preferred
The Micro tube has a volume capacity 0.5 mL. It has a diameter of 10.8 mm and a length of 44 mm. The tube has a conical base with skirted base. This product can be centrifuged up to 20000 x g. This is sold in a case of 1000.
Catalog No. NC0856993 Supplier Andwin Scientific Supplier No. 72730003
The Micro tube has a volume capacity 0.5 mL. It has a diameter of 10.8 mm and a length of 44 mm. The tube has a conical base with skirted base. This product can be centrifuged up to 20000 x g. This is sold in a case of 1000.