Learn More
Covaris Inc microTUBE AFA Fiber Pre-Slit Snap-Cap 6x16mm, 25/pk
SDP
Catalog No. NC9871616
Encompass
6 x 16mm Round bottom glass tube, AFA fiber, and pre-slit snap-cap system. For use with S holder 500114 or E rack 500111.
Catalog No. NC9871616 Supplier Covaris Inc Supplier No. 520045
6 x 16mm Round bottom glass tube, AFA fiber, and pre-slit snap-cap system. For use with S holder 500114 or E rack 500111.