Learn More
Covaris Inc MICROTUBE AFA SCREWCAP 25/PK
SDP
Catalog No. NC0380760
Encompass
microTUBE AFA Fiber Screw-Cap 6x16mm. Legacy product designed to process 50ul samples using the M220 Focused-ultrasonicator and M220 Holder microTUBE 500301. Please ensure that you have M220 Holder microTUBE (500301) before ordering this product.
Catalog No. NC0380760 Supplier Covaris Inc Supplier No. 520096
microTUBE AFA Fiber Screw-Cap 6x16mm. Legacy product designed to process 50ul samples using the M220 Focused-ultrasonicator and M220 Holder microTUBE 500301. Please ensure that you have M220 Holder microTUBE (500301) before ordering this product.