Learn More
Research Products International Corp Micro-Tube Cap Locks, Lift Tab Style, 1.5ml - 2.0ml Tubes, Assorted Colors, 100 per Package
SDP
Catalog No. 50550173
Encompass
Unique Micro-Tube Cap Locks securely seal tubes. Ideal for high temperature incubation, freezing and thawing procedures. Locks prevent caps from opening during temperature and pressure changes. Tab design allows tubes to be lifted easily in and out of boiling water. Provides added security from accidental opening of caps when working with hazardous samples.
Catalog No. 50-550-173 Supplier Research Products International Corp Supplier No. 145063
Unique Micro-Tube Cap Locks securely seal tubes. Ideal for high temperature incubation, freezing and thawing procedures. Locks prevent caps from opening during temperature and pressure changes. Tab design allows tubes to be lifted easily in and out of boiling water. Provides added security from accidental opening of caps when working with hazardous samples.