Learn More
Waters Corp Microtube Domino, 5 mL, Holds 15 Tubes, Automation Compatible
Catalog No. NC3797529 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This 5 mL microtube domino is designed to hold a variety of mid-volume microtubes in various formats. Its design maintains attached caps of certain tubes in a vertical position to prevent automation collisions, making it suitable for seamless automation of laboratory workflows. It can hold up to 15 tubes and is equipped with QR codes for critical labware information and ARUCO codes for spatial positioning.
  • Designed to hold a variety of mid-volume microtubes in various formats.
  • Maintains attached caps of certain tubes in a vertical position to prevent automation collisions.
  • Holds up to 15 tubes.
  • Functions as a machine-readable consumable holder for seamless automation.
  • Equipped with QR codes for critical labware information.
  • Includes ARUCO codes for spatial positioning.
Catalog No. NC3797529 Supplier Waters Corp Supplier No. 186010202
This 5 mL microtube domino is designed to hold a variety of mid-volume microtubes in various formats. Its design maintains attached caps of certain tubes in a vertical position to prevent automation collisions, making it suitable for seamless automation of laboratory workflows. It can hold up to 15 tubes and is equipped with QR codes for critical labware information and ARUCO codes for spatial positioning.
  • Designed to hold a variety of mid-volume microtubes in various formats.
  • Maintains attached caps of certain tubes in a vertical position to prevent automation collisions.
  • Holds up to 15 tubes.
  • Functions as a machine-readable consumable holder for seamless automation.
  • Equipped with QR codes for critical labware information.
  • Includes ARUCO codes for spatial positioning.