Learn More
Research Products International Corp Colored Micro-Tube Freezer Box, Purple
SDP
Catalog No. 501367864
Encompass
Cardboard Freezer Boxes have a unique color band design and ample white space for quick, easy labeling and sample identification. Boxes can hold an 81 or 100 capacity cell divider (sold separately).
Fits most standard size freezer racks.
Catalog No. 50-136-7864 Supplier Research Products International Corp Supplier No. 181058PR
Cardboard Freezer Boxes have a unique color band design and ample white space for quick, easy labeling and sample identification. Boxes can hold an 81 or 100 capacity cell divider (sold separately).
Fits most standard size freezer racks.