Learn More
Research Products International Corp Micro-Tube Ice Rack, Polypropylene, for 1.5 ml Tubes, 50 Tube Capacity
SDP
Catalog No. 50550408
Encompass
Ideal rack for processing heat sensitive samples. Two-piece design allows convenient access for filling with ice. Heavy duty polypropylene provides excellent durability for repeated use.
Catalog No. 50-550-408 Supplier Research Products International Corp Supplier No. H18905
Ideal rack for processing heat sensitive samples. Two-piece design allows convenient access for filling with ice. Heavy duty polypropylene provides excellent durability for repeated use.