Learn More
Research Products International Corp MICRO-TUBE ICE RACK, POLYSTYRENE, HOLDS 0.5ML TUBES,105 CAPACITY
SDP
Catalog No. 50212958
Encompass
Ideal rack for temperature stabilized benchtop procedures or storage. Two piece design allows convenient access for filling with water or ice. High-impact polystyrene provides excellent durability for repeated use.
Catalog No. 50-212-958 Supplier Research Products International Corp Supplier No. H18901
Ideal rack for temperature stabilized benchtop procedures or storage. Two piece design allows convenient access for filling with water or ice. High-impact polystyrene provides excellent durability for repeated use.