Learn More
Biologix Technologies Inc BLX-038B Desktop Microtube Labels — BLUE, 350 labels/roll, 2 rolls/pk
Catalog No. NC2852168
Encompass
Same labels as BLX-038 with a blue stripe.Circle top and a rectangle for the side. These Microtube labels fit all standard 1.5 – 2.0 ml Microcentrifuge tubes. Waterproof and freezer safe.2 rolls/pk350 labels/roll
Catalog No. NC2852168 Supplier Biologix Technologies Inc Supplier No. BLX038B
Same labels as BLX-038 with a blue stripe.Circle top and a rectangle for the side. These Microtube labels fit all standard 1.5 – 2.0 ml Microcentrifuge tubes. Waterproof and freezer safe.2 rolls/pk350 labels/roll