Learn More
USA Scientific Inc Pestle for 0.5 mL microcentrifuge tubes
Catalog No. NC9719656
Encompass
Pestle for 0.5 mL microcentrifuge tubes, polypropylene, 10 bags of 10 (100 pestles)
Catalog No. NC9719656 Supplier USA Scientific Inc Supplier No. 14054390
Pestle for 0.5 mL microcentrifuge tubes, polypropylene, 10 bags of 10 (100 pestles)