Learn More
Waters Corp Microtube Rack Domino, 0.2 mL, 24-Position, Andrew+ Compatible
Catalog No. NC3831116 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
  • Holds up to 24, 0.2 mL microtubes.
  • Compatible with the Andrew+ pipetting robot.
  • Rack is compatible with the microplate gripper tool.
  • Features machine-readable consumable holders.
  • Equipped with QR codes housing critical labware information.
  • Equipped with ARUCO codes for spatial positioning.
Catalog No. NC3831116 Supplier Waters Corp Supplier No. 186010303
  • Holds up to 24, 0.2 mL microtubes.
  • Compatible with the Andrew+ pipetting robot.
  • Rack is compatible with the microplate gripper tool.
  • Features machine-readable consumable holders.
  • Equipped with QR codes housing critical labware information.
  • Equipped with ARUCO codes for spatial positioning.