Learn More
Research Products International Corp Cardboard Micro-Tube Storage Box Set with Cell Partition, 81 Tube Capacity
SDP
Catalog No. 50550219
Encompass
RPI's cardboard storage box is made of heavy duty cardboard and is ideal for freezer and refrigerator storage. Boxes contain a cardboard cell partition to hold popular size micro-tubes (0.2 - 2.0 ml).
Helps maximize space
Boxes have a moisture resistant covering
Catalog No. 50-550-219 Supplier Research Products International Corp Supplier No. 181032
RPI's cardboard storage box is made of heavy duty cardboard and is ideal for freezer and refrigerator storage. Boxes contain a cardboard cell partition to hold popular size micro-tubes (0.2 - 2.0 ml).
Helps maximize space
Boxes have a moisture resistant covering