Learn More
Research Products International Corp Cardboard Micro-Tube Storage Box Set with Cell Partition, 100 Tube Capacity, 48 Boxes per Case
SDP
Catalog No. 50998393
Encompass
RPI's cardboard storage box is made of heavy duty cardboard and is ideal for freezer and refrigerator storage. Boxes contain a cardboard cell partition to hold popular size micro-tubes (0.2 - 2.0 ml).
Helps maximize space
Boxes have a moisture resistant covering
Catalog No. 50-998-393 Supplier Research Products International Corp Supplier No. 181028CS
RPI's cardboard storage box is made of heavy duty cardboard and is ideal for freezer and refrigerator storage. Boxes contain a cardboard cell partition to hold popular size micro-tubes (0.2 - 2.0 ml).
Helps maximize space
Boxes have a moisture resistant covering