Learn More
Research Products International Corp Acrylic Micro-Centrifuge Tube Rack for 0.5 ml Tubes, 54 Capacity
SDP
Catalog No. 50212362
Encompass
These acrylic racks are available for 0.5, 1.5, 1.7, and 2.0ml micro-tubes. Two-tier design supports tube to allow easy opening and closing of hinged lids. Racks are constructed of heavy duty transparent acrylic.
Catalog No. 50-212-362 Supplier Research Products International Corp Supplier No. 146154
These acrylic racks are available for 0.5, 1.5, 1.7, and 2.0ml micro-tubes. Two-tier design supports tube to allow easy opening and closing of hinged lids. Racks are constructed of heavy duty transparent acrylic.