Learn More
Andwin Scientific MICROTUBE TOUGH-SPOTS YELLOW
SDP
Catalog No. NC9155015
Encompass_Preferred
The Tough-Spots labels on a roll are specially designed to be used for 2.0 mL tubes. The labels are resistant against chemicals and temperatures from -196C to 80C. This product can be put on several surfaces like most plastics, glass, vinyl, and metals. Any marking instrument can also be utilized to write on these labels. This product has a diameter of 1/2". It is sold in a pack of 1000 yellow labels.
Catalog No. NC9155015 Supplier Andwin Scientific Supplier No. TSPOTS50Y
The Tough-Spots labels on a roll are specially designed to be used for 2.0 mL tubes. The labels are resistant against chemicals and temperatures from -196C to 80C. This product can be put on several surfaces like most plastics, glass, vinyl, and metals. Any marking instrument can also be utilized to write on these labels. This product has a diameter of 1/2". It is sold in a pack of 1000 yellow labels.