Learn More
Research Products International Corp Acrylic Micro-Centrifuge Tube Rack for 1.5 ml and 2.0 ml Tubes, 40 Tube Capacity
SDP
Catalog No. 50550195
Encompass
These convenient racks are made of clear acrylic for clarity and durability.  Accommodates micro-tubes from 1.5 ml to 2.0 ml.
Catalog No. 50-550-195 Supplier Research Products International Corp Supplier No. 146100
These convenient racks are made of clear acrylic for clarity and durability.  Accommodates micro-tubes from 1.5 ml to 2.0 ml.